Learn More
Azenta US Inc 1.9ml Tri-coded Tube, 48-format, External Thread
Supplier:  Azenta US Inc 657643GS
Encompass
This item has a minimum order quantity of 10 per supplier requirements
The 1.9ml Tri-coded Tube, 48-format, External Thread offers unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.
Catalog No. NC2193025