Learn More
Enzo Life Sciences 1alpha,25-Dihydroxyvitamin D3, (50µg), CAS Number: 32222-06-3
SDP
Supplier:  Enzo Life Sciences BMLDM2000050
Encompass_Preferred
Ultra-high purity. 1α,25-Dihydroxyvitamin D.50=16 nM). Up-regulates RANKL level and down-regulates OPG level. Alternative Name: 1alpha,25-Dihydroxycholecalciferol, Calcitriol. Formula: C27H44O3. MW: 416.6. CAS Number: 32222-06-3. Purity: ≥99% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO, 100% ethanol,  ethyl acetate, tetrahydrofuran, chloroform or acetone. Evaporative loss of solvent may occur during storage and handling of the dissolved product. If necessary, exact. Concentration may be redetermined spectrophotometrically. Long Term Storage: -80°C
Catalog No. NC0011602