Learn More
Medchemexpress LLC 1-(a-D-ribofuranosyl)uracil | 3258-07-9 | MFCD17430297 | 97.0% | 244.20 g/mol | C9H12N2O6 | 250 MG

Supplier: Medchemexpress LLC HYW128112250MG
1-(a-D-ribofuranosyl)uracil is a purine nucleoside analog supplied as a solid for research use. It exhibits antitumor activity by inhibiting DNA synthesis and inducing apoptosis, and is provided with stated purity and storage recommendations to support laboratory handling and long-term storage.
- Molecular formula: C9H12N2O6.
- CAS number: 3258-07-9.
- Purity: 97.0%.
- Molecular weight: 244.20 g/mol.
- Appearance: solid, white to off-white.
- Available pack sizes include 50 MG, 100 MG, 250 MG, 500 MG, and 1 G; solution format 10 mM in DMSO.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.