Learn More
Medchemexpress LLC 1-Adamantylcarbinyl iodide | 51849-10-6 | 95.0% | 276.16 | 5 MG

Supplier: Medchemexpress LLC HY424045MG
1-Adamantylcarbinyl iodide is a drug intermediate used for the synthesis of various active compounds. It is for research use only.
- Drug intermediate
- Solid appearance
- White to off-white color
- Store at 4°C, protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.