Learn More
Medchemexpress LLC 1-adamantylmethyl 4-((2,5-dihydroxybenzyl)amino)benzoate | 241127-79-7 | 99.5% | 407.50 g/mol | C25H29NO4 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12357825MG
Encompass_Preferred
NSC689857 is a small-molecule research compound reported to inhibit epidermal growth factor receptor (EGFR) and the SCF Skp2 complex, with reported IC50 values of ~36 μM for Skp2-Cks1 and ~30 μM for inhibition of p27 ubiquitylation. It is supplied as a white to off-white solid (molecular formula C25H29NO4; molecular weight 407.50 g/mol) and is intended for laboratory research use only.
  • EGFR and Skp2 inhibitory activity (IC50 ≈36 μM)
  • Inhibits p27 ubiquitylation (IC50 ≈30 μM)
  • High reported purity (~99.5%)
  • Solid, white to off-white appearance
  • Stable when stored as recommended (powder -20°C long term; solvent -80°C short term)
  • For laboratory research use only
Catalog No. 50-002-37513