Learn More
Medchemexpress LLC 1-Amino-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48-hexadecaoxahenpentacontan-51-oic acid | 2922656-52-6 | 95.0% | C35H71NO18 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14017910MG
Encompass_Preferred
Amino-PEG16-acid is an amine-terminated polyethylene glycol (PEG16) linker designed for use in the chemical synthesis of proteolysis targeting chimeras (PROTACs) and other bifunctional conjugates. It provides a hydrophilic, flexible spacer with reactive termini suitable for orthogonal coupling strategies and is supplied as a research-grade reagent with a reported purity of 95%.
  • Amine-terminated PEG16 linker useful for PROTAC synthesis.
  • Hydrophilic and flexible spacer to improve solubility and reduce steric hindrance.
  • Terminal carboxylic acid and primary amine for orthogonal conjugation chemistries.
  • Reported purity 95.0% suitable for research applications.
  • Molecular weight ~793.93 Da; formula C35H71NO18.
Catalog No. 50-002-60195