Learn More
Medchemexpress LLC 1-Azabicyclo[2.2.2]octan-3-one, 2,2-bis(hydroxymethyl)- | 5608-24-2 | 98.0% | 185.22 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19980A
Encompass_Preferred
PRIMA-1 is a mutant p53 reactivator that restores the sensitivity of TP53 mutant-type thyroid cancer cells to the histone methylation inhibitor 3-Deazaneplanocin A.
  • Reactivates mutant p53, restoring sensitivity in TP53 mutant-type thyroid cancer cells.
  • Significantly suppresses lung adenocarcinoma formation in mice.
  • Shows in vitro activity with IC50 values ranging from 35 to 75 μM in various cell lines.
  • Demonstrates in vivo activity, suppressing lung adenocarcinoma formation.
Catalog No. 50-004-02732