Learn More
Medchemexpress LLC HY-59090 2mg Medchemexpress, 1-Azakenpaullone CAS:676596-65-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY590902MG
Encompass_Preferred
Medchemexpress, HY-59090 2mg 1-Azakenpaullone CAS:676596-65-9 1-Azakenpaullone (1-Akp) is a highly selective and ATP-competitive inhibitor of glycogen synthase kinase-3 β (GSK-3β) , with an IC 50 value of 18 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2873