Learn More
Medchemexpress LLC 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-, chloride (1:1) | 528-58-5 | MFCD00017582 | 322.70 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0499
Encompass_Preferred
Cyanidin Chloride (IdB 1027) is a subclass of anthocyanin that displays antioxidant and anti-carcinogenesis properties. It is known to inhibit osteoclast formation, hydroxyapatite resorption, and receptor activator of NF-κB ligand (RANKL)-induced osteoclast marker gene expression.
  • Antioxidant properties.
  • Anti-carcinogenesis properties.
  • Inhibits osteoclast formation.
  • Inhibits hydroxyapatite resorption.
  • Inhibits RANKL-induced osteoclast marker gene expression.
Catalog No. 50-003-89912