Learn More
Medchemexpress LLC (1-benzyl-1H-indol-3-yl)methanol | 60941-76-6 | MFCD01312953 | 99.7% | 237.30 g·mol⁻¹ | C16H15NO | 10 MG

Supplier: Medchemexpress LLC HYN1015910MG
1-Benzyl-I3C ((1-benzyl-1H-indol-3-yl)methanol) is an indole-3-carbinol derivative reported as a NEDD4-1 inhibitor for biochemical and cellular research into ubiquitination and cancer-related signaling. Provided as a solid, white to off-white material with defined purity and solubility properties, it is intended for small-scale experimental applications.
- High purity (99.7%).
- Molecular weight 237.30 g·mol⁻¹.
- CAS number 60941-76-6.
- Soluble in DMSO at 100 mg/mL.
- Storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).
- Supplied in research-scale quantities, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.