Learn More
Medchemexpress LLC 1-benzyl-3-(4-methoxyanilino)-4-phenylpyrrole-2,5-dione | 264206-85-1 | 99.2% | 384.43 g/mol | C24H20N2O3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12340250MG
Encompass_Preferred
GSK3987 is a pan LXRα/β agonist research compound used to study cholesterol metabolism and lipogenesis. It increases expression of ABCA1 and SREBP-1c, promotes cellular cholesterol efflux, and can induce triglyceride accumulation. Supplied for laboratory research use only, the compound is supported by a certificate of analysis and product documentation.
Catalog No. 50-002-38506