Learn More
Medchemexpress LLC 1-benzyl-3-cyclohexyl-1-(piperidin-4-ylmethyl)urea hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100347100MG
Encompass_Preferred
SRI-011381 hydrochloride is an orally active small-molecule agonist of the transforming growth factor-β (TGF-β)/Smad signaling pathway. Supplied as a purified hydrochloride salt for research use, it has been reported to exhibit neuroprotective effects and to modulate TGF-β-dependent cellular responses in vitro and in vivo. The material is provided as a solid with high analytical purity suitable for preclinical experiments.
  • Orally active TGF-β/Smad signaling agonist.
  • Exhibits reported neuroprotective effects in preclinical studies.
  • Supplied as a hydrochloride solid suitable for formulation and dosing.
  • High purity for research applications.
  • Molecular weight 365.94 g/mol; CAS 2070014-88-7.
Catalog No. 50-002-14169