Learn More
Medchemexpress LLC 1-benzyl-3-cyclohexyl-1-(piperidin-4-ylmethyl)urea; hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 500 MG
SDP
Supplier:  Medchemexpress LLC HY100347A500MG
Encompass_Preferred
SRI-011381 hydrochloride is a small-molecule TGF-β/Smad signaling agonist (C381) that targets lysosomes and shows neuroprotective effects in preclinical models. Supplied as the hydrochloride salt for research use, it has been used to study lysosomal function and neurodegeneration mechanisms.
  • Orally active TGF-β/Smad signaling agonist.
  • Targets lysosomes to promote acidification and proteolysis.
  • Demonstrated neuroprotective effects in preclinical studies.
  • Hydrochloride salt form for improved solubility and handling.
  • Available in multiple solid sizes and as DMSO solutions for assay-ready use.
Catalog No. 50-002-14007