Learn More
Medchemexpress LLC 1-Benzyl-3-phenylthi 5g
SDP
Supplier:  Medchemexpress LLC HYW1119995G
Encompass_Preferred
1-Benzyl-3-phenylthiourea is a sensor material for detecting nerve agents and related simulants and has biosensor activity 1-Benzyl-3-phenylthiourea can be used to increase the sensitivity of sensors to nerve agents and enhance detection capabilities The reaction characteristics of 1-Benzyl-3-phenylthiourea are similar to those of nerve agents and their simulants making it show good results in detection[1]
Catalog No. 50-002-06517