Learn More
Medchemexpress LLC 1-Benzyl-4-hydroxy-N,N,2-trimethyl-1H-benzo[d]imidazole-6-carboxamide | 1640981-20-9 | 99.6% | C18H19N3O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY47848
Encompass_Preferred
1-Benzyl-4-hydroxy-N,N,2-trimethyl-1H-benzo[d]imidazole-6-carboxamide is a drug intermediate for the synthesis of various active compounds. This product boasts a high purity of 99.6% and is stable under recommended storage conditions. It is suitable for use as a laboratory chemical and in the manufacture of substances.
  • High purity of 99.6%
  • Stable for up to 3 years at room temperature
  • Stable for up to 2 years in solvent at -80°C
  • Stable for up to 1 year in solvent at -20°C
  • Suitable for synthesis of various active compounds
  • Identified for laboratory and manufacturing uses
Catalog No. 50-003-59028