Learn More
Medchemexpress LLC 1-Benzyl-4-hydroxy-N,N,2-trimethyl-1H-benzo[d]imidazole-6-carboxamide | 1640981-20-9 | 99.6% | C18H19N3O2 | 5 G

Supplier: Medchemexpress LLC HY478485G
1-Benzyl-4-hydroxy-N,N,2-trimethyl-1H-benzo[d]imidazole-6-carboxamide is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is also suitable for general laboratory chemical applications and the manufacture of substances.
- Used as a drug intermediate
- Pathway: Others
- Suitable for laboratory chemical applications
- Appearance: Off-white to pale purple solid
- Purity: 99.6% (HPLC)
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.