Learn More
Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | MFCD00006192 | 189.25 | 1000 G
Supplier: Medchemexpress LLC HYW0078221000G
1-Benzyl-4-piperidone is a versatile chemical compound primarily used in organic synthesis and medicinal chemistry. It serves as a key intermediate in the production of various pharmaceutical compounds. This product is suitable for laboratory use and experienced personnel.
- Suitable for organic synthesis
- Applicable in medicinal chemistry research
- Store long-term at 2-8°C in pure form for optimal stability
- Shipping at room temperature if less than 2 weeks