Learn More
Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | MFCD00006763 | 189.25 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW007822
Encompass_Preferred
1-Benzyl-4-piperidone is a versatile intermediate in organic synthesis, serving as a building block for various pharmaceuticals and organic compounds. It is frequently employed in the synthesis of piperidine derivatives, which are prevalent in medicinal chemistry due to their diverse biological activities. This compound acts as a key precursor in the production of analgesics, antipsychotics, and other central nervous system agents.
  • Versatile intermediate in organic synthesis.
  • Building block for pharmaceuticals and organic compounds.
  • Used in the synthesis of piperidine derivatives.
  • Precursor for analgesics, antipsychotics, and central nervous system agents.
  • Carbonyl group facilitates reductive amination, Grignard additions, and condensation reactions.
  • Benzyl group enables further modifications.
Catalog No. 50-004-50002