Learn More
Cayman Chemical 1-BenzylImIdazole 5g
SDP
Supplier:  Cayman Chemical 705105 G
Encompass_Preferred
1-Benzylimidazole is a selective inhibitor of thromboxane synthase. It reduces TXB2 levels and increases blood flow in ischemia-reperfusion injury of rat brain.{2001}
Catalog No. 50-289-7024