Learn More
Medchemexpress LLC 1-(Benzyloxy)-2-iodobenzene | 142523-69-1 | 99.2% | C13H11IO | 1 G
SDP
Supplier:  Medchemexpress LLC HY761281G
Encompass_Preferred
1-(Benzyloxy)-2-iodobenzene is a drug intermediate used for the synthesis of various active compounds. It is a colorless to light yellow liquid with a purity of 99.23% and a molecular weight of 310.13. It is intended for research use only.
  • Drug intermediate for synthesis
  • Colorless to light yellow liquid
  • High purity for research applications
Catalog No. 50-004-34166