Learn More
Medchemexpress LLC 1-(Benzyloxy)-2-iodobenzene | 142523-69-1 | 99.2% | C13H11IO | 100 MG

Supplier: Medchemexpress LLC HY76128100MG
1-(Benzyloxy)-2-iodobenzene is a drug intermediate for the synthesis of various active compounds.
- Molecular weight: 310.13
- Appearance: Liquid
- Color: Colorless to light yellow
- Standard shipping: Room temperature in continental US; may vary elsewhere
- Storage conditions: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
- CAS number: 142523-69-1
- Purity: 99.23%
- Chemical formula: C13H11IO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.