Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(Benzyloxy)-2-iodobenzene | 142523-69-1 | 99.2% | C13H11IO | 5 G
SDP

Supplier:  Medchemexpress LLC HY761285G

Encompass_Preferred

1-(Benzyloxy)-2-iodobenzene is a drug intermediate utilized for the synthesis of various active compounds.

  • Used for synthesizing active compounds
  • Molecular weight: 310.13
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-004-34115


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.