Learn More
Medchemexpress LLC 1-(Benzyloxy)-2-iodobenzene | 142523-69-1 | MFCD14648490 | 99.2% | C13H11IO | 500 MG

Supplier: Medchemexpress LLC HY76128500MG
1-(Benzyloxy)-2-iodobenzene is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only.
- Purity of 99.2%
- Molecular weight of 310.13
- Appears as a colorless to light yellow liquid
- Store at 4°C, protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.