Learn More
Medchemexpress LLC 1-(Bis(dimethylamino)methylene)-1H-benzotriazolium hexafluorophosphate(1-) 3-oxide | 94790-37-1 | MFCD00075445 | 98.0% | 379.24 g/mol | C11H16F6N5OP | 25g
SDP
Supplier:  Medchemexpress LLC HYY091225G
Encompass_Preferred
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]
Catalog No. 50-002-69782