Learn More
Medchemexpress LLC 1-Boc-2-(S)-pyrrolidinemethanol | 69610-40-8 | 99.9% | C10H19NO3 | 25 G

Supplier: Medchemexpress LLC 25GHY78926
1-Boc-2-(S)-pyrrolidinemethanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has a molecular weight of 201.26 and appears as a white to yellow solid. This product is for research use only.
- Can be used as a biochemical reagent
- Suitable as a biological material
- Acts as an organic compound
- Ideal for life science related research
- Molecular weight: 201.26
- Appearance: White to yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.