Learn More
Medchemexpress LLC 1-Boc-3-Aminopyrrolidine | 186550-13-0 | MFCD01861220 | 99.9% | 186.25 | 5 G

Supplier: Medchemexpress LLC HY400915G
1-Boc-3-Aminopyrrolidine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Appearance: Solid-Liquid Mixture
- Color: Colorless to off-white
- Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)
- Shipping: Room temperature in continental US; may vary elsewhere
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.