Learn More
Medchemexpress LLC 1-Boc-3-(hydroxymethyl)pyrrolidine | 114214-69-6 | 98.5% | C10H19NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY602185G
Encompass_Preferred
1-Boc-3-(hydroxymethyl)pyrrolidine is an intermediate that can be used to synthesize a FGFR1/2/3/4 inhibitor and is applicable in cancer research.
  • Can be used to synthesize a FGFR1/2/3/4 inhibitor
  • Applicable in cancer research
  • Categorized as an intermediate
Catalog No. 50-004-34741