Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-4-(2-hydroxyethyl)piperazine | 77279-24-4 | 99.6% | 230.30 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY20797

Encompass_Preferred

Boc-Piperazine-OH is a PROTAC linker. It is a chemical compound with a molecular weight of 230.30 and a high purity of 99.55%. It appears as a solid below 39°C and a liquid above 39°C, typically colorless to light yellow. This product is intended for research use only.

  • Functions as a PROTAC linker
  • Appearance is solid below 39°C, liquid above 39°C
  • Color is colorless to light yellow
  • Purity of 99.55%
  • Molecular weight of 230.30
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-02882


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.