Learn More
Medchemexpress LLC 1-BOC-4-(2-Hydroxyethyl)piperazine-d4 | 1219802-10-4 | 99% | 234.33 | 100 MG
Supplier: Medchemexpress LLC HY20797S100MG
1-BOC-4-(2-Hydroxyethyl)piperazine-d4 is the deuterium labeled form of 1-BOC-4-(2-Hydroxyethyl)piperazine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
- This compound can be used as a tracer.
- This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.