Learn More
Medchemexpress LLC 1-Boc-4-(4-aminobutyl)piperazine | 745048-07-1 | MFCD04038543 | 95.0% | 257.37 g/mol | C13H27N3O2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW045078
1-Boc-4-(4-aminobutyl)piperazine is a Boc-protected piperazine derivative used as a chemical intermediate in medicinal chemistry for the synthesis of bioactive compounds. It is typically supplied in laboratory-scale quantities and is used where a protected primary amine is required for subsequent coupling or deprotection steps.
- Boc-protected piperazine scaffold for selective functionalization.
- Suitable as a building block in drug synthesis.
- Typical purity reported at 95%.
- Molecular weight about 257.37 g/mol and formula C13H27N3O2.
- Physical appearance: oil, colorless to light yellow.
- Recommended storage: protect from light; cool storage advised for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.