Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-4-(4-aminobutyl)piperazine | 745048-07-1 | MFCD04038543 | 95.0% | 257.37 g/mol | C13H27N3O2 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0450785G

Encompass_Preferred

1-Boc-4-(4-aminobutyl)piperazine is a Boc-protected piperazine derivative used as a drug intermediate in medicinal chemistry and organic synthesis.

  • High purity (≥95.0%).
  • Molecular weight 257.37 g/mol and formula C13H27N3O2.
  • Oily liquid, colorless to light yellow.
  • Available in multiple package sizes, including 5 g.
  • Store at 4°C and protect from light; in solvent stability: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-10164


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.