Learn More
Medchemexpress LLC 1-Boc-4-carboxymethyl piperazine | 156478-71-6 | 97.0% | C11H20N2O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HY750055G
Encompass_Preferred
1-Boc-4-carboxymethyl piperazine is a chemical compound that serves as a PROTAC linker, a crucial component in targeted protein degradation. This linker is specifically utilized in the synthesis of various PROTACs, including PROTAC IRAK4 degrader-12.
  • Functions as a PROTAC linker
  • Essential for the synthesis of PROTACs
  • Exhibits 97.0% purity
  • Appears as an off-white to gray solid
  • Stable for 3 years at room temperature
Catalog No. 50-004-34781