Learn More
Medchemexpress LLC 1-Boc-homopiperazine | 112275-50-0 | 99.9% | 200.28 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY40181
Encompass_Preferred
1-Boc-homopiperazine is a drug intermediate that can be used for the synthesis of bacterial FtsZ inhibitors. It has a molecular weight of 200.28, formula C10H20N2O2, and appears as a liquid, colorless to light yellow. It is for research use only.
  • Drug intermediate.
  • Can be used for the synthesis of bacterial FtsZ inhibitors.
  • High purity (99.87% / 99.92%).
  • Available in multiple quantities for various research needs.
  • Detailed documentation available.
  • Solubility information provided for in vitro and in vivo use.
  • Related to research on antibacterial agents and bacterial cell division inhibitors.
Catalog No. 50-004-03134