Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-pyrazole-4-boronic acid pinacol ester | 552846-17-0 | 294.16 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW00323310MM/1ML

Encompass_Preferred

1-Boc-pyrazole-4-boronic acid pinacol ester is a biochemical reagent. It can be utilized as a biological material or an organic compound for life science-related research. This product is intended for research use only and not for sale to patients.

  • Appearance: White to off-white solid
  • Purity: 98.0%
  • Soluble in DMSO (200 mg/mL)
  • Suitable for life science research

Catalog No. 50-004-45384


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.