Learn More
Medchemexpress LLC 1-bromo-3,4-difluorobenzene-d3 | 1219799-14-0 | >=98.0% | 196.0073 g·mol⁻¹ | C6D3BrF2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW015079S50MG
Encompass_Preferred
1-Bromo-3,4-difluorobenzene-d3 is a deuterium-labeled aromatic compound supplied as a neat material for use as a stable isotope reference and as a labeled intermediate in synthesis. It provides high deuterium incorporation and chemical purity suitable for analytical quantitation and research workflows.
  • Deuterium-labeled compound (C6D3BrF2) for isotopic tracing and quantitation.
  • High deuterium enrichment, ≥98 atom % D.
  • High chemical purity, ≥98%.
  • Molecular weight ~196.01 g·mol⁻¹ for accurate mass calculations.
  • Suitable as a reference standard and labeled intermediate in synthesis.
  • Supplied neat in small-format packages for laboratory use.
Catalog No. 50-003-02653