Learn More
Medchemexpress LLC 1-bromo-3,4-difluorobenzene-d3 | 1219799-14-0 | 98.6% | 196.01 | C6D3BrF2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015079S5MG
Encompass_Preferred
1-Bromo-3,4-difluorobenzene-d3 is a deuterium-labeled aromatic reagent supplied as a high-purity, isotopically enriched liquid for use as a stable-isotope reference or synthetic building block in research chemistry. It is characterized by GC purity and specified storage conditions for long-term stability.
  • Deuterium-labeled aromatic reagent suitable for analytical and synthetic use.
  • High chemical purity as measured by gas chromatography.
  • High isotopic enrichment for accurate mass and tracer studies.
  • Colorless to light yellow liquid, easy-to-handle neat material.
  • Recommended storage at low temperatures for long-term stability.
Catalog No. 50-003-03662