Learn More
Medchemexpress LLC 1-bromo-3,4-difluorobenzene-d3 | 1219799-14-0 | 98.6% | 196.01 g/mol | C6D3BrF2 | 10 MG

Supplier: Medchemexpress LLC HYW015079S10MG
1-Bromo-3,4-difluorobenzene-d3 is the deuterium-labeled analogue of 1-bromo-3,4-difluorobenzene, intended for use as an isotopic standard, tracer, or synthetic building block. Supplied as a neat liquid, the material is characterized by QC documentation and recommended storage conditions to preserve isotopic integrity and purity.
- Deuterium-labeled (d3) isotopologue suitable for tracer and standard applications.
- High isotopic enrichment (~98.5%) and high GC purity (~98.6%).
- Molecular formula C6D3BrF2 and molecular weight 196.01 g/mol.
- Neat liquid physical form for direct use in synthesis workflows.
- Certificate of analysis and safety data sheet available for quality assurance.
- Recommended storage conditions documented to maintain stability and shelf life.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.