Learn More
Medchemexpress LLC 1-Bromo-3,5-bis(trifluoromethyl)benzene | 328-70-1 | 99.94% | C8H3BrF6 | 1 KG
SDP
Supplier:  Medchemexpress LLC HY750881KG
Encompass_Preferred
1-Bromo-3,5-bis(trifluoromethyl)benzene is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It is also utilized in the preparation of the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion.
  • Functions as a stabilizing counterion for electrophilic organic and organometallic cations
  • Suitable for life science related research
  • Can be used as a biological material or organic compound
Catalog No. 50-004-34387