Learn More
Medchemexpress LLC 1-Bromo-3-chlorobenzene-d4 | 1643577-43-8 | 195.48 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW010004S1G
Encompass_Preferred
1-Bromo-3-chlorobenzene-d4 (3-Chlorobromobenzene-d4) is a deuterium labeled 1-Bromo-3-chlorobenzene. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
Catalog No. 50-003-66423