Learn More
Medchemexpress LLC 1-Bromo-3-chlorobenzene-d4 | 1643577-43-8 | 99.4% | 195.48 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW010004S500MG
Encompass_Preferred
1-Bromo-3-chlorobenzene-d4 (3-Chlorobromobenzene-d4) is a deuterium labeled 1-Bromo-3-chlorobenzene. It is intended for research use only and not for patient use. This compound can be utilized as a tracer or as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including deuterium, are incorporated into drug molecules primarily as tracers for quantitation during drug development, with potential to influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Features stable heavy isotopes for drug development research
Catalog No. 50-004-53940