Learn More
Medchemexpress LLC 1-Bromo-3-iodobenzene | 591-18-4 | 99.5% | 282.90 | 1 KG
SDP
Supplier:  Medchemexpress LLC HY321491KG
Encompass_Preferred
1-Bromo-3-iodobenzene is a biochemical reagent and an important organic synthetic intermediate. It can be used for the synthesis of other active compounds and acts as a dihaloarene substrate in palladium-catalyzed three-component tandem reactions.
  • Biochemical reagent
  • Important organic synthetic intermediate
  • Can be used for the synthesis of other active compounds
  • Acts as a dihaloarene substrate in palladium-catalyzed three-component tandem reactions
  • Enables one-pot synthesis of unsymmetrical biaryl thioglycosides with thiosugars and arylboronic acids
  • Facilitates selective formation of S-C and C-C bonds for efficient transformation
Catalog No. 50-004-01088