Learn More
Medchemexpress LLC 1-Bromo-3-iodobenzene-d4 | 2363787-31-7 | 286.93 | 1 G
SDP
Supplier:  Medchemexpress LLC HY32149S1G
Encompass_Preferred
1-Bromo-3-iodobenzene-d4 is the deuterium labeled form of 1-Bromo-3-iodobenzene, which is a biochemical reagent and an important organic synthetic intermediate. This compound can be utilized for the synthesis of other active compounds. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Isotope-labeled compound
  • Biochemical assay reagent
  • Drug intermediate
  • Used for the synthesis of other active compounds
Catalog No. 50-004-00504