Learn More
Medchemexpress LLC 1-bromo-3-methoxybenzene-d3 | 873928-72-4 | MFCD12760295 | 190.05 g·mol⁻¹ | C7H4D3BrO | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW016152S100MG
Encompass_Preferred
1-Bromo-3-methoxybenzene-d3 is the deuterium-labeled analogue of 1-bromo-3-methoxybenzene (CAS 873928-72-4) used as an isotopic tracer for analytical and metabolic studies. Three deuterium atoms replace the methoxy hydrogens, producing a defined mass shift that enables accurate tracking and quantitation in mass spectrometry workflows.
  • Isotopically labeled for precise mass spectrometry quantitation.
  • Suitable as an internal standard in metabolic tracing and pharmacokinetic studies.
  • Deuterium substitution provides a clear mass shift for unambiguous identification.
  • Compatible with common analytical techniques, including LC-MS and GC-MS.
Catalog No. 50-003-03691