Learn More
Medchemexpress LLC 1-bromo-4-phenoxybenzene-d5 | 93951-83-8 | 98.8% | 254.13 | C12H4D5BrO | 10 MG
SDP
Supplier:  Medchemexpress LLC HY76595S10MG
Encompass_Preferred
1-Bromo-4-phenoxybenzene-d5 is a deuterium-labeled analogue of 1-bromo-4-phenoxybenzene provided as an isotopically labeled reference standard for analytical and research applications. It is designed for use as an internal standard in mass spectrometry, isotope tracing, and quantitative assays requiring deuterium labeling. CAS 93951-83-8, molecular formula C12H4D5BrO, exact mass 254.13, purity 98.8%.
  • Deuterium-labeled (d5) isotopic standard suitable for mass spectrometry
  • High reported purity enhances analytical accuracy
  • Suitable as an internal standard for quantitative assays
  • Available in small milligram quantities for analytical workflows
Catalog No. 50-002-98623