Learn More
Medchemexpress LLC 1-Bromododecane-d3 | 204259-68-7 | 98.0% | 252.25 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008758S110MG
Encompass_Preferred
1-Bromododecane-d3 is the deuterium-labeled version of 1-Bromododecane. It is intended for research purposes only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for research applications during the drug development process
Catalog No. 50-004-51263