Learn More
Medchemexpress LLC 1-bromooctane-d4 | 1219803-37-8 | 197.15 g/mol | C8H13D4Br | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW032015S310MG
Encompass_Preferred
1-Bromooctane-d4 is the deuterium-labeled analogue of 1-bromooctane used as an isotope-labeled standard for analytical chemistry and drug-development studies. It replaces four hydrogen atoms with deuterium on the octyl chain, enabling mass-based discrimination and accurate quantitation in mass spectrometry workflows.
  • Contains four deuterium atoms replacing hydrogens on the octyl chain.
  • Designed as an isotope-labeled standard for quantitative mass spectrometry.
  • Useful as a tracer in metabolic and ADME studies.
  • Provided in a pure form suitable for analytical applications.
  • Supplied in research-scale quantities for method development.
Catalog No. 50-002-99462