Learn More
Medchemexpress LLC 1-bromoundecane-1,1,2,2-d4 | 1219802-82-0 | 98.0% | 1 MG

Supplier: Medchemexpress LLC HY122426S21MG
1-bromoundecane-d4 is a deuterium-labeled alkyl bromide supplied for use as an isotopic tracer or internal standard in analytical chemistry methods such as NMR, GC-MS, and LC-MS.
- Deuterium-labeled analogue of 1-bromoundecane (d4).
- CAS number: 1219802-82-0.
- Molecular formula: C11H19D4Br; molecular weight: 239.23.
- Intended for research use as an analytical standard or tracer.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.