Learn More
Medchemexpress LLC 1-butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide | 623580-02-9 | MFCD14156017 | ≥97.0% | 436.43 g·mol-1 | C12H22F6N2O4S2 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW12770125G
Encompass_Preferred
1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide is an ionic liquid composed of a 1-butyl-1-methylpiperidinium cation paired with the bis(trifluoromethylsulfonyl)imide (NTf2) anion. Supplied for research use, it functions as a high-purity, low-volatility solvent or electrolyte for electrochemical studies, battery research, and synthetic applications.
  • High thermal and electrochemical stability.
  • Low volatility and negligible vapor pressure.
  • Good ionic conductivity for electrolyte applications.
  • Supplied with specified purity (≥97.0%).
  • Suitable as solvent or supporting electrolyte in electrochemistry and battery research.
  • Available in laboratory-scale packaging for research workflows.
Catalog No. 50-002-82655