Learn More
Medchemexpress LLC 1-Butyl-1-methylpyrrolidinium chloride | 479500-35-1 | MFCD08458922 | ≥98.0% | 177.72 g·mol-1 | C9H20ClN | 100 G

Supplier: Medchemexpress LLC HYW016638100G
1-Butyl-1-methylpyrrolidinium chloride is a quaternary ammonium chloride salt provided for laboratory research as a high-purity ionic liquid. It serves as a solvent and reagent in synthetic and materials chemistry and should be handled according to the safety data sheet and institutional procedures.
- High purity: ≥98.0%.
- Molecular formula: C9H20ClN.
- Molecular weight: 177.72 g·mol-1.
- Function: ionic liquid, solvent, and phase-transfer agent.
- Physical form: typically supplied as a liquid/solid salt for bench use.
- Use restrictions: for research use only; not for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.