Learn More
Medchemexpress LLC 1-Butynylbenzene-d5 | 1219803-38-9 | 135.22 g/mol | C10H5D5 | 10 MG

Supplier: Medchemexpress LLC HYW142175S10MG
1-Butynylbenzene-d5 is the deuterium-labeled analogue of 1-butynylbenzene provided as an isotope-labeled reagent for research use. It contains an alkyne functional group and is suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC), making it useful in analytical workflows and synthetic click chemistry applications.
- Deuterium-labeled for tracer and isotope studies.
- Contains an alkyne group compatible with CuAAC click chemistry.
- Useful as an internal standard in analytical quantitation.
- Molecular formula C10H5D5 and molecular weight 135.22 g/mol.
- Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.